Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Nitril Gloves,S7.5,White,Cuff:3.54mil,Palm:5.91mil,Finger Tip:7.09mil ,Sterile 1 * 100 items
Catalog # ANSEBNMS-75
Supplier:
Ansell
Expand image
Nitril Gloves,S7.5,White,Cuff:3.54mil,Palm:5.91mil,Finger Tip:7.09mil ,Sterile 1 * 100 items
Catalog # ANSEBNMS-75
Supplier:
Ansell