Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid Protein, His tag (E.Coli) 1 * 1 mg
Catalog # ICNA08720502
Supplier:
MP Biomedicals
Expand image
SARS-CoV-2 Nucleocapsid Protein, His tag (E.Coli) 1 * 1 mg
Catalog # ICNA08720502
Supplier:
MP Biomedicals