Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pc,cryobox,2 ,81-place 1-2ml,133x133x52mm,unhinged,assorted 1 * 24 items
Catalog # IBSAPC2X8UNHA
Supplier:
IBS Stock Items
Expand image
pc,cryobox,2 ,81-place 1-2ml,133x133x52mm,unhinged,assorted 1 * 24 items
Catalog # IBSAPC2X8UNHA
Supplier:
IBS Stock Items