Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Human Her2, His Tag (HE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 100 µG
ACRBHE2-H5225100UG
:
ACROBIOSYSTEMS
Human Her2, His Tag (HE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 100 µG
ACRBHE2-H5225100UG
:
ACROBIOSYSTEMS