Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
pH REFERENCE STANDARD BUFFER COLORLESS 1 * 4 L
pH REFERENCE STANDARD BUFFER COLORLESS 1 * 4 L
Print
pH REFERENCE STANDARD BUFFER COLORLESS 1 * 4 L
Catalog # BDHB50260-4A
Supplier:
BDH US PRODUCED
pH REFERENCE STANDARD BUFFER COLORLESS 1 * 4 L
Catalog # BDHB50260-4A
Supplier:
BDH US PRODUCED
Expand image
Return to Top
pH REFERENCE STANDARD BUFFER COLORLESS 1 * 4 L
Add to cart
Price
$289.00
Log in
for availability
Unit of Measure
1 * 4 L
-
+
Add to cart
Return Policy
Product Details & Documents