Order Entry
Singapore
ContactUsLinkComponent
Spore Suspensions, stoppered and crimp sealed, spores suspended in 40% ethanol solution packed 10ml in glass vial 1 * 1 items
Catalog # XXXXSSGE/8
Supplier:  Chemicals
Spore Suspensions, stoppered and crimp sealed, spores suspended in 40% ethanol solution packed 10ml in glass vial 1 * 1 items
Catalog # XXXXSSGE/8
Supplier:  Chemicals