Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ortho-Phosphoric Acid AR,ACS 5L 1 * 5 L
Catalog # RANKO0057
Supplier:
RANKEM
Certificates
Expand image
Ortho-Phosphoric Acid AR,ACS 5L 1 * 5 L
Catalog # RANKO0057
Supplier:
RANKEM
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation