Order Entry
Singapore
ContactUsLinkComponent
Side access rack system to complete the SU780UE/XLE upright freezer 4.75inches boxes with 36 place dividers for 15mL tubes (20 rack s, 160 box capacity) BOXES and DIVIDERS INCLUDED 1 * 1 items
Catalog # GLCL780-RS5-RACK
Supplier:  Stirling Ultracold
Side access rack system to complete the SU780UE/XLE upright freezer 4.75inches boxes with 36 place dividers for 15mL tubes (20 rack s, 160 box capacity) BOXES and DIVIDERS INCLUDED 1 * 1 items
Catalog # GLCL780-RS5-RACK
Supplier:  Stirling Ultracold