Order Entry
Singapore
ContactUsLinkComponent
Side access rack system to complete the SU780UE/XLE upright freezer 3inches boxes (30 racks, 450 box capacity) RACKS and SHELVES O NLY 1 * 1 items
Catalog # GLCL780-RKIT2
Supplier:  Stirling Ultracold
Side access rack system to complete the SU780UE/XLE upright freezer 3inches boxes (30 racks, 450 box capacity) RACKS and SHELVES O NLY 1 * 1 items
Catalog # GLCL780-RKIT2
Supplier:  Stirling Ultracold