Order Entry
Singapore
ContactUsLinkComponent
Side access rack for the SU105UE undercounter freezer 2inches boxes (12 box capacity, 3 high x 4 deep) RACK ONLY 1 * 1 items
Catalog # GLCL105-R-342
Supplier:  Stirling Ultracold
Side access rack for the SU105UE undercounter freezer 2inches boxes (12 box capacity, 3 high x 4 deep) RACK ONLY 1 * 1 items
Catalog # GLCL105-R-342
Supplier:  Stirling Ultracold