Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Fully Autoclavable Variable Volume Micropipette 0.5-10µl 1 * 1 items
FINEAV-100
:
ACCUMAX
Fully Autoclavable Variable Volume Micropipette 0.5-10µl 1 * 1 items
FINEAV-100
:
ACCUMAX