Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
200µl Universal Grad tip, Sterile, Low retention (Individually Wrapped) 1 * 400 items
Catalog # FINEAT-200-S-L-W
Supplier:
ACCUMAX
Expand image
200µl Universal Grad tip, Sterile, Low retention (Individually Wrapped) 1 * 400 items
Catalog # FINEAT-200-S-L-W
Supplier:
ACCUMAX