Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Class A Glass, 0.030 tolerance, individually wrapped 1 * 1 items
Catalog # EISCCH0559H
Supplier:
EISCO
Expand image
Class A Glass, 0.030 tolerance, individually wrapped 1 * 1 items
Catalog # EISCCH0559H
Supplier:
EISCO