$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Class A Glass, 0.030 tolerance, individually wrapped 1 * 1 items
EISCCH0559G
:
EISCO
Class A Glass, 0.030 tolerance, individually wrapped 1 * 1 items
EISCCH0559G
:
EISCO