Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Storage bags, static dissipative, 24×36 , 4 mil Blue 1 * 100 items
Catalog # VWRMNP24X36BL4M
Supplier:
VWR Products
Expand image
Storage bags, static dissipative, 24×36 , 4 mil Blue 1 * 100 items
Catalog # VWRMNP24X36BL4M
Supplier:
VWR Products