Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE BAG ANTI-STATIC 15A 18IN 4MLBLUE 1 * 100 items
Catalog # VWRMNP0250103150
Supplier:
VWR Products
Expand image
STORAGE BAG ANTI-STATIC 15A 18IN 4MLBLUE 1 * 100 items
Catalog # VWRMNP0250103150
Supplier:
VWR Products