Order Entry
Singapore
ContactUsLinkComponent
High intensive Polymide tape 3 inch core 5mm x 33m thickness 0.06mm High Temperature resistance 260 degreec (Qualify) 1 * 1 Roll
Catalog # IBSAJDK19ERISETAPE
Supplier:  IBS Stock Items
High intensive Polymide tape 3 inch core 5mm x 33m thickness 0.06mm High Temperature resistance 260 degreec (Qualify) 1 * 1 Roll
Catalog # IBSAJDK19ERISETAPE
Supplier:  IBS Stock Items