Order Entry
Singapore
ContactUsLinkComponent
Low IgG Grade bovine albumin. The albumin is Chromatographically extracted from NZ bovine plasma, under Quality Systems that are ISO 9001 Certified and audited to cGMP principles. 1 * 1 kg
Catalog # ICNA0219989791
Supplier:  MP Biomedicals
Low IgG Grade bovine albumin. The albumin is Chromatographically extracted from NZ bovine plasma, under Quality Systems that are ISO 9001 Certified and audited to cGMP principles. 1 * 1 kg
Catalog # ICNA0219989791
Supplier:  MP Biomedicals