Order Entry
Singapore
ContactUsLinkComponent
Microbiological Grade bovine albumin. The albumin is Chromatographically extracted from NZ bovine plasma, under Quality Systems that are ISO 9001 Certified and audited to cGMP principles. 1 * 500 g
Catalog # ICNA0218062090
Supplier:  MP Biomedicals
Microbiological Grade bovine albumin. The albumin is Chromatographically extracted from NZ bovine plasma, under Quality Systems that are ISO 9001 Certified and audited to cGMP principles. 1 * 500 g
Catalog # ICNA0218062090
Supplier:  MP Biomedicals