Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FASTCELLULAR SENESCENCE DETECTION KIT 1 * 1 KIT
Catalog # ICNA092690301
Supplier:
MP Biomedicals
Expand image
FASTCELLULAR SENESCENCE DETECTION KIT 1 * 1 KIT
Catalog # ICNA092690301
Supplier:
MP Biomedicals