Order Entry
Singapore
Orders LinkContactUsLinkComponent
Freezing media

Freezing media

Freezing medium, cryopreservation-medium, with DMSO, with FBS, 10 mL
Catalog # PANBP07-90010
Marketsource
Supplier:   PAN BIOTECH
Freezing media
Catalog # PANBP07-90010
Supplier:  PAN BIOTECH
Marketsource
CAS Number:  
Freezing media

Freezing medium, cryopreservation-medium, with DMSO, with FBS, 10 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 10 mL
Product Details & Documents

Freezing medium, cryopreservation-medium for the cryoconservation of cells. The medium is based on DMEM, supplemented with a mix of fetal bovine serum and DMSO.

  • Stored at −20 °C

This composition guarantees a high survival rate and excellent cell growth after thawing.

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    Freezing medium, cryopreservation-medium, with DMSO, with FBS

Specifications