Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
IMDM, without glucose, without L-glutamine, with 25 mM HEPES, with 3,024 g/L NaHCO 1 * 500 mL
PANBP04-20158
:
PAN BIOTECH
IMDM, without glucose, without L-glutamine, with 25 mM HEPES, with 3,024 g/L NaHCO 1 * 500 mL
PANBP04-20158
:
PAN BIOTECH