Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Star Magnetic Stirring Bars, PTFE-encapsulated magnetic core (Alnico 5), universal chemical resistance. 1 * 1 items
Catalog # BOHLC352-06
Supplier:
Bohlender
Expand image
Star Magnetic Stirring Bars, PTFE-encapsulated magnetic core (Alnico 5), universal chemical resistance. 1 * 1 items
Catalog # BOHLC352-06
Supplier:
Bohlender