Order Entry
Singapore
ContactUsLinkComponent
Chamfer Magnetic Stirring Bars, PTFE-encapsulated magnetic core (Alnico 5), crosswise chamfered top side, flat bottom, universal che mical resistance. 1 * 1 items
Catalog # BOHLC356-02
Supplier:  Bohlender
undefined
Chamfer Magnetic Stirring Bars, PTFE-encapsulated magnetic core (Alnico 5), crosswise chamfered top side, flat bottom, universal che mical resistance. 1 * 1 items
Catalog # BOHLC356-02
Supplier:  Bohlender