$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
MP FILTRATION SYSTEM 10L 1 * 1 items
KIML953976-1003
:
KIMBLEKONTES
MP FILTRATION SYSTEM 10L 1 * 1 items
KIML953976-1003
:
KIMBLEKONTES