Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CRUCIBLE GOOCH LOW FORM FRITTED F 30ML 1 * 12 items
Catalog # KIML28260-303
Supplier:
KIMBLEKONTES
Expand image
CRUCIBLE GOOCH LOW FORM FRITTED F 30ML 1 * 12 items
Catalog # KIML28260-303
Supplier:
KIMBLEKONTES