Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
THERM ONLY NON MERCURY 10-25ML 14-38C 1 * 1 items
KIML15123N-2538
:
KIMBLEKONTES
THERM ONLY NON MERCURY 10-25ML 14-38C 1 * 1 items
KIML15123N-2538
:
KIMBLEKONTES