Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REACTION TUBE W/DRAFT SHIELD AM/PH 50ML 1 * 1 items
Catalog # KIML479470-0050
Supplier:
KIMBLEKONTES
Expand image
REACTION TUBE W/DRAFT SHIELD AM/PH 50ML 1 * 1 items
Catalog # KIML479470-0050
Supplier:
KIMBLEKONTES