Order Entry
Singapore
ContactUsLinkComponent
Top plate: 210 x 210 mm, Width X Depth: 275 x 350 mm, Height: 245mm, Net weight: 24 kg, Electric supply: 220v 50hz 1 * 1 items
Catalog # IBSAMBF-200NC
Supplier:  IBS Stock Items
Top plate: 210 x 210 mm, Width X Depth: 275 x 350 mm, Height: 245mm, Net weight: 24 kg, Electric supply: 220v 50hz 1 * 1 items
Catalog # IBSAMBF-200NC
Supplier:  IBS Stock Items