Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISIONHT C18 HIGHLOAD 150X4.6MM 5um 1 * 1 items
Catalog # MAIS5151970
Supplier:
DR MAISCH
Expand image
VISIONHT C18 HIGHLOAD 150X4.6MM 5um 1 * 1 items
Catalog # MAIS5151970
Supplier:
DR MAISCH