Order Entry
Singapore
ContactUsLinkComponent
Freezer CryoCube F740h ULT 5 inner compartments door handle left air-cooled with chart recorder LN2-backup system 230V/50Hz EU-plug 1 * 1 items
Catalog # NEWBF740345031
Supplier:  NEW BRUNSWICK
Freezer CryoCube F740h ULT 5 inner compartments door handle left air-cooled with chart recorder LN2-backup system 230V/50Hz EU-plug 1 * 1 items
Catalog # NEWBF740345031
Supplier:  NEW BRUNSWICK