Order Entry
Singapore
ContactUsLinkComponent
Freezer CryoCube F740h ULT 5 inner compartments door handle right air-cooled with chart recorder 230V/50Hz UK-plug 1 * 1 items
  NEWBF740341042
Freezer CryoCube F740h ULT 5 inner compartments door handle right air-cooled with chart recorder 230V/50Hz UK-plug 1 * 1 items
  NEWBF740341042