Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Freezer CryoCube F740h ULT 3 inner compartments door handle right air-cooled with chart recorder 230V/50Hz EU-plug 1 * 1 items
NEWBF740341021
:
NEW BRUNSWICK
Freezer CryoCube F740h ULT 3 inner compartments door handle right air-cooled with chart recorder 230V/50Hz EU-plug 1 * 1 items
NEWBF740341021
:
NEW BRUNSWICK