Order Entry
Singapore
ContactUsLinkComponent
Freezer CryoCube F740h ULT 5 inner compartments door handle right air-cooled 230V/50Hz EU-plug 1 * 1 items
Catalog # NEWBF740340041
Supplier:  NEW BRUNSWICK
Freezer CryoCube F740h ULT 5 inner compartments door handle right air-cooled 230V/50Hz EU-plug 1 * 1 items
Catalog # NEWBF740340041
Supplier:  NEW BRUNSWICK