Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
BUFFER, PRECISION REFERENCE STANDARD, PH 1 * 1 items
BUFFER, PRECISION REFERENCE STANDARD, PH 1 * 1 items
Print
BUFFER, PRECISION REFERENCE STANDARD, PH 1 * 1 items
Catalog # RICCR1618300-500G
Supplier:
Ricca Chemical
BUFFER, PRECISION REFERENCE STANDARD, PH 1 * 1 items
Catalog # RICCR1618300-500G
Supplier:
Ricca Chemical
Expand image
Return to Top
BUFFER, PRECISION REFERENCE STANDARD, PH 1 * 1 items
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Return Policy
Product Details & Documents