Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Modification kit CO2/FPI 3-sensor 1 * 1 KIT
Catalog # BIND9501-0046
Supplier:
Binder
Expand image
Modification kit CO2/FPI 3-sensor 1 * 1 KIT
Catalog # BIND9501-0046
Supplier:
Binder