Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MAGNESIUM NITRATE (2,0% MG) IN NITRIC ACID 5% MATRIX MODIFIER SOLUTION 1 * 50 mL
Catalog # RICCRMMG20KN-50N
Supplier:
Ricca Chemical
Expand image
MAGNESIUM NITRATE (2,0% MG) IN NITRIC ACID 5% MATRIX MODIFIER SOLUTION 1 * 50 mL
Catalog # RICCRMMG20KN-50N
Supplier:
Ricca Chemical