Order Entry
Singapore
ContactUsLinkComponent
Low Flow Ultrasonic Benchtop Flow Meter (1/16in ID), flow range 2-100mL/min, console unit with 24VDC power supply and Freq and mA ou tpu
Catalog # PENDFM-US-LF-C
Supplier:  PENDOTECH
Low Flow Ultrasonic Benchtop Flow Meter (1/16in ID), flow range 2-100mL/min, console unit with 24VDC power supply and Freq and mA ou tpu
Catalog # PENDFM-US-LF-C
Supplier:  PENDOTECH