Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
0.5M BUFFERS BRITTON-ROBINSON BUFFER, PH 1.98, ADJUSTED TO 0.5 M IONIC STRENGTH 1 * 500 mL
Catalog # RICCR1156200-500A
Supplier:
Ricca Chemical
Expand image
0.5M BUFFERS BRITTON-ROBINSON BUFFER, PH 1.98, ADJUSTED TO 0.5 M IONIC STRENGTH 1 * 500 mL
Catalog # RICCR1156200-500A
Supplier:
Ricca Chemical