Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
0.1M BUFFERS BRITTON-ROBINSON BUFFER, PH 11.96, ADJUSTED TO 0.1 M IONIC STRENGTH 1 * 500 mL
Catalog # RICCR1155200-500A
Supplier:
Ricca Chemical
Expand image
0.1M BUFFERS BRITTON-ROBINSON BUFFER, PH 11.96, ADJUSTED TO 0.1 M IONIC STRENGTH 1 * 500 mL
Catalog # RICCR1155200-500A
Supplier:
Ricca Chemical