MILLIPAK-40, 200 CM² UNIT, WITH DURAPORE MEMBRANE IN PVDF LOW PROTEIN ADSORPTION, 0.1 ΜM, INLET FITTINGS AND CANNEL OUTPUT FOR 6MM P IPE, EXIT WITH BELL FILLING, STERILIZED BY GAMMA RAYS, BOX OF 2 1 * 2 items
MILLIPAK-40, 200 CM² UNIT, WITH DURAPORE MEMBRANE IN PVDF LOW PROTEIN ADSORPTION, 0.1 ΜM, INLET FITTINGS AND CANNEL OUTPUT FOR 6MM P IPE, EXIT WITH BELL FILLING, STERILIZED BY GAMMA RAYS, BOX OF 2 1 * 2 items