Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY CALIBRATION STANDARD – OF 200 1 * 500 mL
Catalog # CANN9727-E27.016
Supplier:
CANNON INSTRUMENT
Expand image
VISCOSITY CALIBRATION STANDARD – OF 200 1 * 500 mL
Catalog # CANN9727-E27.016
Supplier:
CANNON INSTRUMENT