Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IMDM, w: stab. Glutamine, w: 25 mM Hepes, w/o: Phenol red, null
Catalog # PANBP04-20451
Supplier:
PAN BIOTECH
Expand image
IMDM, w: stab. Glutamine, w: 25 mM Hepes, w/o: Phenol red, null
Catalog # PANBP04-20451
Supplier:
PAN BIOTECH