Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IMDM, w: stable glutamine, w: 25 mM HEPES, w: 3,024 g / L NaHCO3, 500ml 1 * 500 mL
Catalog # PANBP04-20450
Supplier:
PAN BIOTECH
Expand image
IMDM, w: stable glutamine, w: 25 mM HEPES, w: 3,024 g / L NaHCO3, 500ml 1 * 500 mL
Catalog # PANBP04-20450
Supplier:
PAN BIOTECH