Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IMDM, w/o: L-Glutamine, w: 1,0 g/l Glucose, w: 3,024 g/l NaHCO3 1 * 500 mL
Catalog # PANBP04-20259
Supplier:
PAN BIOTECH
Expand image
IMDM, w/o: L-Glutamine, w: 1,0 g/l Glucose, w: 3,024 g/l NaHCO3 1 * 500 mL
Catalog # PANBP04-20259
Supplier:
PAN BIOTECH