Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
DMEM w: 1,0 g/l Glucose, w: L-Glutamine, w/o: Sodium pyruvate, w: 3,7 g/l NaHCO3 1 * 500 mL
PANBP04-01555
:
PAN BIOTECH
DMEM w: 1,0 g/l Glucose, w: L-Glutamine, w/o: Sodium pyruvate, w: 3,7 g/l NaHCO3 1 * 500 mL
PANBP04-01555
:
PAN BIOTECH