Order Entry
Singapore
ContactUsLinkComponent
DMEM, w / o Glucose, w / o L-Glutamine, w / o Sodium pyruvate, w / o HEPES, w 3.7 g / L NaHCO3 1 * 500 mL
Catalog # PANBP04-01548S1
Supplier:  PAN BIOTECH
DMEM, w / o Glucose, w / o L-Glutamine, w / o Sodium pyruvate, w / o HEPES, w 3.7 g / L NaHCO3 1 * 500 mL
Catalog # PANBP04-01548S1
Supplier:  PAN BIOTECH