Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MEM Eagle w: EBSS, w: L-Glutamine, w: NEAA, w/o: NaHCO3 1 * 50 L
Catalog # PANBP03-2950
Supplier:
PAN BIOTECH
Expand image
MEM Eagle w: EBSS, w: L-Glutamine, w: NEAA, w/o: NaHCO3 1 * 50 L
Catalog # PANBP03-2950
Supplier:
PAN BIOTECH