Order Entry
Singapore
ContactUsLinkComponent
NucleoBond Xtra BAC Buffer Set buffer set for the isolation of large constructs, like BACs, PACs, and P1, only applicable with Nucle oBond Xtra BAC kits - Buffer RES-BAC, Buffer LYS-BAC, Buffer NEU-BAC (1000 mL each), RNase A no dangerous goods acco 1 * 1 items
Catalog # MANA740437
Supplier:  MACHEREY-NAGEL
undefined
NucleoBond Xtra BAC Buffer Set buffer set for the isolation of large constructs, like BACs, PACs, and P1, only applicable with Nucle oBond Xtra BAC kits - Buffer RES-BAC, Buffer LYS-BAC, Buffer NEU-BAC (1000 mL each), RNase A no dangerous goods acco 1 * 1 items
Catalog # MANA740437
Supplier:  MACHEREY-NAGEL