Order Entry
Singapore
ContactUsLinkComponent
Compact photometer PF-3 Pool incl. test kits VISOCOLOR ECO Chlorine 2 free and total (liquid), pH 6.0-8.2, Cyanuric acid, Alkalinity TA, with calibration cuvette, 4 empty tubes, beaker, 5 mL syringe, 3 batteries, manual, booklet VISOCOLOR ECO Test 1 * 1 SET
Catalog # MANA934118
Supplier:  MACHEREY-NAGEL
undefined
Compact photometer PF-3 Pool incl. test kits VISOCOLOR ECO Chlorine 2 free and total (liquid), pH 6.0-8.2, Cyanuric acid, Alkalinity TA, with calibration cuvette, 4 empty tubes, beaker, 5 mL syringe, 3 batteries, manual, booklet VISOCOLOR ECO Test 1 * 1 SET
Catalog # MANA934118
Supplier:  MACHEREY-NAGEL
Related Information